Anti-IL18 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 74801]

Supplier: R & D Systems
IC646P
102884-674EA 566.89 USD
102884-674
Anti-IL18 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 74801]
Antibodies
Type: Primary
Antigen: IL18
Clonality: Monoclonal
Clone: 74801
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR