Anti-EBI3 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 607201]

Supplier: R & D Systems
IC6456A
102884-670EA 633.42 USD
102884-670
Anti-EBI3 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 607201]
Antibodies
Type: Primary
Antigen: EBI3
Clonality: Monoclonal
Clone: 607201
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR