Anti-TWIST1 Sheep Polyclonal Antibody (Fluorescein)

Supplier: R & D Systems
IC6230F
220018-471EA 404.92 USD
220018-471
Anti-TWIST1 Sheep Polyclonal Antibody (Fluorescein)
Antibodies
Type: Primary
Antigen: TWIST1
Clonality: Polyclonal
Clone:
Conjugation: Fluorescein
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR