Anti-LAMP2 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 743320]

Supplier: R & D Systems
IC6228G
102884-662EA 485.91 USD
102884-662
Anti-LAMP2 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 743320]
Antibodies
Type: Primary
Antigen: LAMP2
Clonality: Monoclonal
Clone: 743320
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR