Anti-EOMES Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 644730]

Supplier: R & D Systems
IC6166G
102884-652EA 608.83 USD
102884-652
Anti-EOMES Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 644730]
Antibodies
Type: Primary
Antigen: EOMES
Clonality: Monoclonal
Clone: 644730
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR