Anti-EOMES Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 644730]

Supplier: R & D Systems
IC6166A
102884-650EA 642.09 USD
102884-650
Anti-EOMES Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 644730]
Antibodies
Type: Primary
Antigen: EOMES
Clonality: Monoclonal
Clone: 644730
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR