Anti-CSF2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 6804]

Supplier: R & D Systems
IC615P
220018-469EA 513.38 USD
220018-469
Anti-CSF2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 6804]
Antibodies
Type: Primary
Antigen: CSF2
Clonality: Monoclonal
Clone: 6804
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR