Anti-RORC Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 600214]

Supplier: R & D Systems
IC6109C
220018-468EA 584.25 USD
220018-468
Anti-RORC Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 600214]
Antibodies
Type: Primary
Antigen: RORC
Clonality: Monoclonal
Clone: 600214
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR