Anti-IDO1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 700838]

Supplier: R & D Systems
IC6030P
102884-326EA 582.8 USD
102884-326
Anti-IDO1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 700838]
Antibodies
Type: Primary
Antigen: IDO1
Clonality: Monoclonal
Clone: 700838
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR