Anti-IDO1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 700838]

Supplier: R & D Systems
IC6030A
102884-646EA 608.83 USD
102884-646
Anti-IDO1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 700838]
Antibodies
Type: Primary
Antigen: IDO1
Clonality: Monoclonal
Clone: 700838
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR