Anti-RORC Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 600380]

Supplier: R & D Systems
IC6006C-025 IC6006C-100
102884-638EA 310.92 USD
102884-638 102884-640
Anti-RORC Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 600380]
Antibodies
Type: Primary
Antigen: RORC
Clonality: Monoclonal
Clone: 600380
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR