Anti-RORC Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 600380]

Supplier: R & D Systems
IC6006A-100 IC6006A-025
102884-636EA 577.02 USD
102884-636 102884-634
Anti-RORC Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 600380]
Antibodies
Type: Primary
Antigen: RORC
Clonality: Monoclonal
Clone: 600380
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR