Anti-AR Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 523339]

Supplier: R & D Systems
IC5876P
102884-626EA 543.75 USD
102884-626
Anti-AR Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 523339]
Antibodies
Type: Primary
Antigen: AR
Clonality: Monoclonal
Clone: 523339
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR