Anti-AR Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 523339]

Supplier: R & D Systems
IC5876A
102884-624EA 608.83 USD
102884-624
Anti-AR Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 523339]
Antibodies
Type: Primary
Antigen: AR
Clonality: Monoclonal
Clone: 523339
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR