Anti-TLR7 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 533707]

Supplier: R & D Systems
IC5875P
220018-457EA 600.15 USD
220018-457
Anti-TLR7 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 533707]
Antibodies
Type: Primary
Antigen: TLR7
Clonality: Monoclonal
Clone: 533707
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR