Anti-ARG1 Sheep Polyclonal Antibody (PE (Phycoerythrin))

Supplier: R & D Systems
IC5868P
102884-618EA 594.37 USD
102884-618
Anti-ARG1 Sheep Polyclonal Antibody (PE (Phycoerythrin))
Antibodies
Type: Primary
Antigen: ARG1
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR