Anti-ARG1 Sheep Polyclonal Antibody (Fluorescein)

Supplier: R & D Systems
IC5868F
102884-616EA 490.25 USD
102884-616
Anti-ARG1 Sheep Polyclonal Antibody (Fluorescein)
Antibodies
Type: Primary
Antigen: ARG1
Clonality: Polyclonal
Clone:
Conjugation: Fluorescein
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR