Anti-CAV1 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 7C8]

Supplier: R & D Systems
IC5736G
102884-586EA 577.02 USD
102884-586
Anti-CAV1 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 7C8]
Antibodies
Type: Primary
Antigen: CAV1
Clonality: Monoclonal
Clone: 7C8
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR