Anti-RPS6 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 522731]

Supplier: R & D Systems
IC5436G
102884-582EA 577.02 USD
102884-582
Anti-RPS6 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 522731]
Antibodies
Type: Primary
Antigen: RPS6
Clonality: Monoclonal
Clone: 522731
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR