Anti-TBX21 Mouse Monoclonal Antibody (Fluorescein) [clone: 525803]

Supplier: R & D Systems
IC5385F
220018-450EA 404.92 USD
220018-450
Anti-TBX21 Mouse Monoclonal Antibody (Fluorescein) [clone: 525803]
Antibodies
Type: Primary
Antigen: TBX21
Clonality: Monoclonal
Clone: 525803
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR