Anti-IL34 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 578416]

Supplier: R & D Systems
IC5265P
102884-580EA 513.38 USD
102884-580
Anti-IL34 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 578416]
Antibodies
Type: Primary
Antigen: IL34
Clonality: Monoclonal
Clone: 578416
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR