Anti-RELA Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 532301]

Supplier: R & D Systems
IC5078P
102884-576EA 549.54 USD
102884-576
Anti-RELA Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 532301]
Antibodies
Type: Primary
Antigen: RELA
Clonality: Monoclonal
Clone: 532301
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR