Anti-LAMP1 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 508921]

Supplier: R & D Systems
IC4800G
102884-562EA 454.09 USD
102884-562
Anti-LAMP1 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 508921]
Antibodies
Type: Primary
Antigen: LAMP1
Clonality: Monoclonal
Clone: 508921
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR