Anti-CCL2 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 123616]

Supplier: R & D Systems
IC479A
102884-560EA 566.89 USD
102884-560
Anti-CCL2 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 123616]
Antibodies
Type: Primary
Antigen: CCL2
Clonality: Monoclonal
Clone: 123616
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR