Anti-SHH Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 171018]

Supplier: R & D Systems
IC4641P
220018-439EA 543.75 USD
220018-439
Anti-SHH Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 171018]
Antibodies
Type: Primary
Antigen: SHH
Clonality: Monoclonal
Clone: 171018
Conjugation: PE (Phycoerythrin)
Epitope: N-Terminal
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR