Anti-FOXP1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 837016]

Supplier: R & D Systems
IC45341A
102884-556EA 577.02 USD
102884-556
Anti-FOXP1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 837016]
Antibodies
Type: Primary
Antigen Symbol: FOXP1
Antigen Name: forkhead box P1
Antigen Synonyms: 12CC4, HSPC215, MFH, QRF1, hFKH1B
Clonality: Monoclonal
Clone: 837016
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR