Anti-CCL3 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 39624]

Supplier: R & D Systems
IC450A
220018-436EA 577.02 USD
220018-436
Anti-CCL3 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 39624]
Antibodies
Type: Primary
Antigen: CCL3
Clonality: Monoclonal
Clone: 39624
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR