Anti-IRF5 Sheep Polyclonal Antibody (Alexa Fluor® 700)

Supplier: R & D Systems
IC4508N
102884-548EA 543.75 USD
102884-548
Anti-IRF5 Sheep Polyclonal Antibody (Alexa Fluor® 700)
Antibodies
Type: Primary
Antigen: IRF5
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 700
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR