Anti-MYHC Mouse Monoclonal Antibody (Fluorescein) [clone: MF20]

Supplier: R & D Systems
IC4470F
220018-435EA 420.83 USD
220018-435
Anti-MYHC Mouse Monoclonal Antibody (Fluorescein) [clone: MF20]
Antibodies
Type: Primary
Antigen: MYHC
Clonality: Monoclonal
Clone: MF20
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR