Anti-GFP Goat Polyclonal Antibody (APC (Allophycocyanin))

Supplier: R & D Systems
IC4240A
102884-538EA 608.83 USD
102884-538
Anti-GFP Goat Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
Type: Primary
Antigen Symbol: GFP
Antigen Name:
Antigen Synonyms: eGFP, GFPuv
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Multiple
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR