Anti-GFP Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 454505]

Supplier: R & D Systems
IC42401A
102884-534EA 644.98 USD
102884-534
Anti-GFP Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 454505]
Antibodies
Type: Primary
Antigen Symbol: GFP
Antigen Name:
Antigen Synonyms: eGFP, GFPuv
Clonality: Monoclonal
Clone: 454505
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Multiple
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR