Anti-P4HB Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 537331]

Supplier: R & D Systems
IC4236G
102884-532EA 561.11 USD
102884-532
Anti-P4HB Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 537331]
Antibodies
Type: Primary
Antigen: P4HB
Clonality: Monoclonal
Clone: 537331
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR