Anti-TNF Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: MP6-XT22]

Supplier: R & D Systems
IC410P
220018-430EA 527.85 USD
220018-430
Anti-TNF Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: MP6-XT22]
Antibodies
Type: Primary
Antigen: TNF
Clonality: Monoclonal
Clone: MP6-XT22
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR