Anti-CXCL9 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 49106]

Supplier: R & D Systems
IC392P
220018-419EA 485.91 USD
220018-419
Anti-CXCL9 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 49106]
Antibodies
Type: Primary
Antigen: CXCL9
Clonality: Monoclonal
Clone: 49106
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR