Anti-RUNX3 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 527327]

Supplier: R & D Systems
IC3765A
220018-415EA 582.8 USD
220018-415
Anti-RUNX3 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 527327]
Antibodies
Type: Primary
Antigen: RUNX3
Clonality: Monoclonal
Clone: 527327
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR