Anti-MYC Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 9E10)]

Supplier: R & D Systems
IC3696P
102884-504EA 600.15 USD
102884-504
Anti-MYC Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 9E10)]
Antibodies
Type: Primary
Antigen: MYC
Clonality: Monoclonal
Clone: 9E10
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR