Anti-MYC Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 9E10)]

Supplier: R & D Systems
IC3696G
102884-500EA 600.15 USD
102884-500
Anti-MYC Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 9E10)]
Antibodies
Type: Primary
Antigen: MYC
Clonality: Monoclonal
Clone: 9E10
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR