Anti-MYC Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 9E10)]

Supplier: R & D Systems
IC3696A
102884-496EA 647.88 USD
102884-496
Anti-MYC Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 9E10)]
Antibodies
Type: Primary
Antigen: MYC
Clonality: Monoclonal
Clone: 9E10
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR