Anti-CX3CL1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 51637]

Supplier: R & D Systems
IC365P
220018-411EA 513.38 USD
220018-411
Anti-CX3CL1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 51637]
Antibodies
Type: Primary
Antigen: CX3CL1
Clonality: Monoclonal
Clone: 51637
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR