Anti-IL33 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 396118]

Supplier: R & D Systems
IC3626P
220018-407EA 537.97 USD
220018-407
Anti-IL33 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 396118]
Antibodies
Type: Primary
Antigen: IL33
Clonality: Monoclonal
Clone: 396118
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR