Anti-PRDM1 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 646702]

Supplier: R & D Systems
IC36081G
102884-316EA 600.15 USD
102884-316
Anti-PRDM1 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 646702]
Antibodies
Type: Primary
Antigen: PRDM1
Clonality: Monoclonal
Clone: 646702
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR