Anti-PRDM1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 646702]

Supplier: R & D Systems
IC36081A
102884-314EA 614.62 USD
102884-314
Anti-PRDM1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 646702]
Antibodies
Type: Primary
Antigen: PRDM1
Clonality: Monoclonal
Clone: 646702
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR