Anti-EGFR Mouse Monoclonal Antibody (Fluorescein) [clone: 338324]

Supplier: R & D Systems
IC3570F
220018-401EA 420.83 USD
220018-401
Anti-EGFR Mouse Monoclonal Antibody (Fluorescein) [clone: 338324]
Antibodies
Type: Primary
Antigen: EGFR
Clonality: Monoclonal
Clone: 338324
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR