Anti-LGALS9 Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 766428]

Supplier: R & D Systems
IC3535G
102884-312EA 519.17 USD
102884-312
Anti-LGALS9 Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 766428]
Antibodies
Type: Primary
Antigen: LGALS9
Clonality: Monoclonal
Clone: 766428
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR