Anti-CXCL12 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 79018]

Supplier: R & D Systems
IC350P
102884-310EA 543.75 USD
102884-310
Anti-CXCL12 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 79018]
Antibodies
Type: Primary
Antigen: CXCL12
Clonality: Monoclonal
Clone: 79018
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR