Anti-CXCL12 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 79018]

Supplier: R & D Systems
IC350C
220018-399EA 577.02 USD
220018-399
Anti-CXCL12 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 79018]
Antibodies
Type: Primary
Antigen: CXCL12
Clonality: Monoclonal
Clone: 79018
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR