Anti-CCL22 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 57203]

Supplier: R & D Systems
IC3361P
220018-396EA 543.75 USD
220018-396
Anti-CCL22 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 57203]
Antibodies
Type: Primary
Antigen: CCL22
Clonality: Monoclonal
Clone: 57203
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR