Anti-SLC27A1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 308420]

Supplier: R & D Systems
IC3304P
220018-391EA 527.85 USD
220018-391
Anti-SLC27A1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 308420]
Antibodies
Type: Primary
Antigen: SLC27A1
Clonality: Monoclonal
Clone: 308420
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR