Anti-FOXP3 Goat Polyclonal Antibody (APC (Allophycocyanin))

Supplier: R & D Systems
IC3240A
220018-388EA 616.06 USD
220018-388
Anti-FOXP3 Goat Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
Type: Primary
Antigen Symbol: FOXP3
Antigen Name: forkhead box P3
Antigen Synonyms: AIID, DIETER, IPEX, JM2, PIDX, XPID
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR