Anti-IL17A Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 41809]

Supplier: R & D Systems
IC317P
102884-476EA 561.11 USD
102884-476
Anti-IL17A Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 41809]
Antibodies
Type: Primary
Antigen: IL17A
Clonality: Monoclonal
Clone: 41809
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR